Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081293.1 | NP_001074762.1 | 7 - 8 | 1127 | 113 | |
GenBank mRNA | AK083674.1 | - | 7 - 8 | 896 | 113 | |
AK087157.1 | - | 7 - 8 | 899 | 113 | ||
AK129150.1 | - | 6 - 7 | 674 | 113 | ||
AK154692.1 | - | 6 - 7 | 801 | 113 | ||
BC067054.1 | - | 7 - 8 | 932 | 30542928 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |