Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700031262.mm.1 |
NM_001159706.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031262.mm.1 |
NM_001159706.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031262.mm.1 |
NM_016770.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031262.mm.1 |
NM_016770.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031262.mm.1 |
TC0700003699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031260.mm.1 |
NM_001159706.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031260.mm.1 |
NM_001159706.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031260.mm.1 |
NM_016770.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031260.mm.1 |
NM_016770.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031260.mm.1 |
TC0700003699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031263.mm.1 |
NM_001159706.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031263.mm.1 |
NM_001159706.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031263.mm.1 |
NM_016770.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031263.mm.1 |
NM_016770.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031263.mm.1 |
TC0700003699.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031261.mm.1 |
NM_001159706.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031261.mm.1 |
NM_001159706.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031261.mm.1 |
NM_016770.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031261.mm.1 |
NM_016770.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031261.mm.1 |
TC0700003699.mm.1 |
MTA TC |