Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008466.5 | NP_032492.1 | 3 - 4 | 354 | 112 | |
GenBank mRNA | AF020772.1 | - | 3 - 4 | 198 | 112 | |
AK011559.1 | - | 3 - 4 | 225 | 112 | ||
AK049373.1 | - | 3 - 4 | 225 | 112 | ||
AK161664.1 | - | 3 - 4 | 282 | 112 | ||
BC026885.1 | - | 3 - 4 | 248 | 3979788 | 112 | |
CJ063714.1 | - | 3 - 4 | 439 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |