Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008991.2 | NP_033017.2 | 7 - 8 | 717 | 123 | |
GenBank mRNA | AK004660.1 | - | 7 - 8 | 698 | 123 | |
AK144060.1 | - | 7 - 8 | 699 | 123 | ||
AK153298.1 | - | 7 - 8 | 675 | 123 | ||
AK198431.1 | - | 4 - 5 | 339 | 123 | ||
BC009119.1 | - | 7 - 8 | 709 | 3154529 | 123 | |
BC054446.1 | - | 7 - 8 | 665 | 6477711 | 123 | |
CB245413.1 | - | 2 - 3 | 165 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |