Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010236.2 | NP_034366.2 | 2 - 3 | 315 | 92 | |
GenBank mRNA | AK041952.1 | - | 2 - 3 | 275 | 92 | |
AK157040.1 | - | 2 - 3 | 276 | 92 | ||
BC005484.1 | - | 2 - 3 | 258 | 3485401 | 92 | |
CA558410.1 | - | 2 - 3 | 168 | 92 | ||
U33557.1 | - | 2 - 3 | 292 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |