Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009267.2 | NP_033293.2 | 3 - 4 | 433 | 120 | |
GenBank mRNA | AK006124.1 | - | 3 - 4 | 431 | 120 | |
BC030164.1 | - | 3 - 4 | 120 | |||
BC106151.1 | - | 3 - 4 | 417 | 4923478 | 120 | |
CK616337.1 | - | 1 - 2 | 120 | |||
CK616968.1 | - | 3 - 4 | 434 | 120 | ||
X00349.1 | - | 3 - 4 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |