Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098638.2 | XP_014954124.1 | 3 - 4 | 761 | 64 | |
XM_027974914.1 | XP_027830715.1 | 3 - 4 | 1968 | 64 | ||
XM_027974916.1 | XP_027830717.1 | 3 - 4 | 1354 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |