Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175453.2 | XP_012030843.2 | 6 - 7 | 860 | 79 | |
XM_012175454.3 | XP_012030844.2 | 6 - 7 | 740 | 79 | ||
XM_012175455.3 | XP_012030845.2 | 6 - 7 | 1662 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |