Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082666.1 | NP_001076135.1 | 3 - 4 | 501 | 69 | |
XM_008266824.2 | XP_008265046.1 | 2 - 3 | 394 | 69 | ||
XM_017346640.1 | XP_017202129.1 | 4 - 5 | 606 | 69 | ||
GenBank mRNA | J05042.1 | - | 3 - 4 | 501 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |