Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093244.4 | XP_001093244.1 | 92 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |