Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194073.3 | NP_001181002.3 | 120 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |