Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194794.3 | NP_001181723.2 | 277 | 69 | ||
GenBank mRNA | JU323404.1 | - | 287 | 69 | ||
JU473240.1 | - | 271 | 69 | |||
JV529498.1 | - | 283 | 69 | |||
JV565037.1 | - | 278 | 69 | |||
JV704655.1 | - | 280 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |