Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017189.2 | NP_058885.1 | 4 - 5 | 507 | 71 | |
XM_006246676.2 | XP_006246738.1 | 4 - 5 | 585 | 71 | ||
GenBank mRNA | AF230645.1 | - | 4 - 5 | 507 | 71 | |
BC088099.1 | - | 4 - 5 | 387 | 7322417 | 71 | |
FQ218583.1 | - | 2 - 3 | 213 | 71 | ||
J02762.1 | - | 4 - 5 | 507 | 71 | ||
M16347.1 | - | 4 - 5 | 486 | 71 | ||
X07636.1 | - | 3 - 4 | 410 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |