Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031349.2 | NP_112639.1 | 1 | 1126 | 62 | |
GenBank mRNA | AB033170.1 | - | 1 | 903 | 62 | |
AF184883.1 | - | 1 | 948 | 62 | ||
BC072494.1 | - | 1 | 1126 | 7097318 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Both primers and probe map within a single exon |