Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127553.2 | NP_001121025.1 | 3 - 4 | 284 | 99 | |
GenBank mRNA | BC166882.1 | - | 3 - 4 | 269 | 6919310 | 99 |
CB783276.1 | - | 3 - 4 | 284 | 99 | ||
FQ217398.1 | - | 3 - 4 | 269 | 99 | ||
FQ217669.1 | - | 3 - 4 | 275 | 99 | ||
FQ217747.1 | - | 1 - 2 | 80 | 99 | ||
FQ217799.1 | - | 3 - 4 | 283 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |