Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105969.1 | NP_001099439.1 | 6 - 7 | 1530 | 60 | |
XM_006250276.2 | XP_006250338.1 | 7 - 8 | 1656 | 60 | ||
XM_006250277.2 | XP_006250339.1 | 5 - 6 | 1445 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |