Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083911.1 | NP_001077380.1 | 1 - 2 | 125 | 68 | |
GenBank mRNA | CB745388.1 | - | 1 - 2 | 277 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |