Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100509.1 | NP_001093979.1 | 8 - 9 | 651 | 69 | |
GenBank mRNA | BC107950.1 | - | 8 - 9 | 572 | 69 | |
FQ213949.1 | - | 8 - 9 | 599 | 69 | ||
FQ214043.1 | - | 8 - 9 | 604 | 69 | ||
FQ227742.1 | - | 8 - 9 | 608 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |