Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012966.1 | NP_037098.1 | 272 | 107 | ||
GenBank mRNA | BC058492.1 | - | 47 | 6887393 | 107 | |
FQ209877.1 | - | 60 | 107 | |||
FQ220992.1 | - | 84 | 107 | |||
FQ222153.1 | - | 59 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |