Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031579.2 | NP_113767.1 | 3 - 4 | 917 | 144 | |
XM_006244670.2 | XP_006244732.1 | 3 - 4 | 799 | 144 | ||
XM_006244671.2 | XP_006244733.1 | 3 - 4 | 782 | 144 | ||
GenBank mRNA | AY062269.1 | - | 2 - 3 | 200 | 144 | |
BC081772.1 | - | 2 - 3 | 547 | 7124031 | 144 | |
BC097307.1 | - | 3 - 4 | 917 | 7446277 | 144 | |
L27843.1 | - | 3 - 4 | 746 | 144 |
Gene Symbol: LOC103693189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766779.1 | XP_008765001.1 | 2 - 3 | 716 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |