Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160079.1 | NP_001153551.1 | 1 | 652 | 110 | |
XM_005667576.3 | XP_005667633.1 | 869 | 110 | |||
GenBank mRNA | AK344174.1 | - | 3 | 810 | 110 | |
FJ875101.1 | - | 1 | 652 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |