Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112425.1 | NP_001105895.1 | 1907 | 108 | ||
GenBank mRNA | U85495.1 | - | 1907 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Zea mays |