Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC75-IV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340610.1 | NP_001329162.1 | 3 - 4 | 515 | 108 | |
NM_001340611.1 | NP_001329163.1 | 2 - 3 | 361 | 108 | ||
NM_116977.3 | NP_192647.2 | 4 - 5 | 607 | 108 | ||
GenBank mRNA | AY585655.1 | - | 4 - 5 | 584 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |