Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCF61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333855.1 | NP_001320817.1 | 5 - 6 | 683 | 75 | |
NM_001333856.1 | NP_001320816.1 | 5 - 6 | 641 | 75 | ||
NM_001333857.1 | NP_001320818.1 | 5 - 6 | 618 | 75 | ||
NM_104650.4 | NP_564745.1 | 5 - 6 | 683 | 75 | ||
GenBank mRNA | AK228076.1 | - | 5 - 6 | 618 | 75 | |
BT006461.1 | - | 5 - 6 | 476 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |