Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114070.4 | NP_566861.1 | 2 - 3 | 1052 | 91 | |
GenBank mRNA | AY084321.1 | - | 2 - 3 | 1013 | 91 | |
AY150426.1 | - | 2 - 3 | 940 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |