Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123762.2 | NP_001117234.1 | 6 - 7 | 867 | 136 | |
NM_100442.5 | NP_563745.1 | 7 - 8 | 1129 | 136 | ||
GenBank mRNA | AY087913.1 | - | 7 - 8 | 843 | 136 | |
BT002049.1 | - | 7 - 8 | 821 | 136 | ||
BT008720.1 | - | 7 - 8 | 724 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |