Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G09010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100772.5 | NP_563833.2 | 6 - 7 | 1006 | 70 | |
GenBank mRNA | AB122060.1 | - | 6 - 7 | 847 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |