Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035579.1 | NP_001030656.1 | 2 | 1547 | 177 | |
NM_001337704.1 | NP_001327376.1 | 2 | 1260 | 177 | ||
NM_001337705.1 | NP_001319492.1 | 3 | 1676 | 177 | ||
NM_001337706.1 | NP_001327372.1 | 2 | 1747 | 177 | ||
NM_001337707.1 | NP_001327375.1 | 2 | 1832 | 177 | ||
NM_001337708.1 | NP_001327373.1 | 2 | 1828 | 177 | ||
NM_001337709.1 | NP_001327374.1 | 3 | 1719 | 177 | ||
NM_001337710.1 | NP_001327371.1 | 2 | 1335 | 177 | ||
NM_111615.4 | NP_566304.1 | 2 | 1631 | 177 | ||
NM_180200.1 | NP_850531.1 | 2 | 1547 | 177 | ||
GenBank mRNA | AY075636.1 | - | 2 | 1487 | 177 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |