Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077020.1 | NP_001070488.1 | 2 - 3 | 137 | 70 | |
GenBank mRNA | BC105572.1 | - | 2 - 3 | 153 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |