Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029230.3 | NP_001024401.1 | 5 - 6 | 526 | 62 | |
NM_001029231.3 | NP_001024402.1 | 5 - 6 | 596 | 62 | ||
NM_001029232.2 | NP_001024403.1 | 5 - 6 | 526 | 62 | ||
NM_001373329.1 | NP_001360660.1 | 5 - 6 | 526 | 62 | ||
NM_001373330.1 | NP_001360659.1 | 5 - 6 | 526 | 62 | ||
GenBank mRNA | AF075245.1 | - | 5 - 6 | 526 | 62 | |
AF117300.1 | - | 5 - 6 | 526 | 62 | ||
AF117301.1 | - | 5 - 6 | 526 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |