Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpdh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129237.4 | NP_001122709.1 | 4 - 5 | 822 | 87 | |
NM_001129238.4 | NP_001122710.1 | 6 - 7 | 985 | 87 | ||
NM_066787.6 | NP_499188.2 | 6 - 7 | 1067 | 87 | ||
NM_181903.7 | NP_871632.1 | 7 - 8 | 1151 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |