Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534705.6 | XP_534705.2 | 3 - 4 | 408 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |