Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427979.1 | XP_022283687.1 | 2 - 3 | 342 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |