Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252190.3 | NP_001239119.1 | 1 | 391 | 95 | |
GenBank mRNA | DN392962.1 | - | 2 | 429 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |