Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497823.1 | XP_003497871.1 | 8 - 9 | 850 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |