Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691309.3 | XP_005691366.2 | 4 - 5 | 887 | 70 | |
XM_018061475.1 | XP_017916964.1 | 4 - 5 | 940 | 70 | ||
GenBank mRNA | JT709836.1 | - | 7 - 8 | 1193 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |