Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058828.1 | XP_017914317.1 | 6 - 7 | 776 | 82 | |
XM_018058829.1 | XP_017914318.1 | 6 - 7 | 827 | 82 | ||
GenBank mRNA | JO631973.2 | - | 6 - 7 | 882 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |