Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056571.1 | XP_017912060.1 | 4 - 5 | 699 | 86 | |
XM_018056572.1 | XP_017912061.1 | 4 - 5 | 736 | 86 | ||
XM_018056573.1 | XP_017912062.1 | 4 - 5 | 652 | 86 | ||
XM_018056574.1 | XP_017912063.1 | 4 - 5 | 691 | 86 | ||
XM_018056575.1 | XP_017912064.1 | 4 - 5 | 507 | 86 | ||
XM_018056576.1 | XP_017912065.1 | 4 - 5 | 554 | 86 | ||
XM_018056577.1 | XP_017912066.1 | 4 - 5 | 561 | 86 | ||
XM_018056578.1 | XP_017912067.1 | 4 - 5 | 490 | 86 | ||
XM_018056579.1 | XP_017912068.1 | 4 - 5 | 701 | 86 | ||
GenBank mRNA | JO427973.1 | - | 4 - 5 | 735 | 86 | |
JO636128.2 | - | 4 - 5 | 490 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |