Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050376.1 | XP_017905865.1 | 7 - 8 | 1066 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |