Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172869.1 | NP_001166340.1 | 156 | 79 | ||
GenBank mRNA | AJ012200.1 | - | 156 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Cavia porcellus |