Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pez | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273198.1 | NP_001260127.1 | 7 - 8 | 4009 | 74 | |
NM_135169.3 | NP_609013.1 | 6 - 7 | 3918 | 74 | ||
GenBank mRNA | AF217284.1 | - | 6 - 7 | 3876 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |