Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmeg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169838.3 | NP_001163309.2 | 8 - 9 | 3114 | 65 | |
NM_001169839.3 | NP_001163310.2 | 8 - 9 | 3235 | 65 | ||
NM_138187.4 | NP_612031.3 | 8 - 9 | 3126 | 65 | ||
NM_167830.4 | NP_728522.2 | 8 - 9 | 3286 | 65 | ||
NM_167831.3 | NP_728523.2 | 9 - 10 | 3323 | 65 | ||
GenBank mRNA | BT044533.1 | - | 8 - 9 | 3194 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |