Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275954.1 | NP_001262883.1 | 5 - 6 | 966 | 98 | |
NM_142926.4 | NP_651183.3 | 5 - 6 | 1347 | 98 | ||
NM_170092.3 | NP_732904.2 | 6 - 7 | 1044 | 98 | ||
NM_170093.3 | NP_732905.2 | 4 - 5 | 1035 | 98 | ||
GenBank mRNA | AF357214.1 | - | 4 - 5 | 891 | 98 | |
AY051436.1 | - | 5 - 6 | 939 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |