Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc17a9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002635.1 | NP_001002635.1 | 12 - 13 | 1459 | 78 | |
GenBank mRNA | BC075944.1 | - | 12 - 13 | 1459 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |