Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myh11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024448.1 | NP_001019619.1 | 19 - 20 | 2514 | 77 | |
XM_021476875.1 | XP_021332550.1 | 20 - 21 | 2567 | 77 | ||
XM_021476876.1 | XP_021332551.1 | 20 - 21 | 2567 | 77 | ||
GenBank mRNA | BC162152.1 | - | 19 - 20 | 2478 | 77 | |
DQ022376.1 | - | 19 - 20 | 2514 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |