Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278002.1 | NP_001264931.1 | 2 - 3 | 300 | 69 | |
NM_001278003.1 | NP_001264932.1 | 2 - 3 | 278 | 69 | ||
GenBank mRNA | BU445269.1 | - | 2 - 3 | 234 | 69 | |
BX930148.1 | - | 2 - 3 | 256 | 69 | ||
BX931864.1 | - | 2 - 3 | 254 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |