Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204689.1 | NP_990020.1 | 2 - 3 | 246 | 111 | |
GenBank mRNA | AB026675.1 | - | 2 - 3 | 246 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |