Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031471.1 | NP_001026642.1 | 3 - 4 | 244 | 108 | |
XM_015275338.2 | XP_015130824.1 | 3 - 4 | 3345 | 108 | ||
GenBank mRNA | AJ720986.1 | - | 3 - 4 | 244 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |