Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204194.1 | NP_989525.1 | 11 - 12 | 1127 | 100 | |
XM_015280909.2 | XP_015136395.1 | 11 - 12 | 1127 | 100 | ||
GenBank mRNA | AF296130.1 | - | 11 - 12 | 1127 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |