Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037279.hg.1 |
TC02002364.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037279.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204709.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204709.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204709.hg.1 |
TC0200014361.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200204705.hg.1 |
TC0200014361.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000345008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000397463.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
ENST00000449209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_001077427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014361.hg.1 |
NM_144586.6 |
Refseq |